Assay Details
| Gene Symbol: DHCR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755653.3 | XP_002755699.1 | 7 - 8 | 1063 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHCR7| Gene Symbol: DHCR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755653.3 | XP_002755699.1 | 7 - 8 | 1063 | 73 | |
Set Membership: |
|