Assay Details
| Gene Symbol: Krt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173033.1 | NP_001166504.1 | 632 | 73 | ||
| GenBank mRNA | EU664998.1 | - | 632 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt12| Gene Symbol: Krt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173033.1 | NP_001166504.1 | 632 | 73 | ||
| GenBank mRNA | EU664998.1 | - | 632 | 73 | ||
Set Membership: |
|