Assay Details
| Gene Symbol: Girdin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274429.1 | NP_001261358.1 | 3 - 4 | 1506 | 95 | |
| NM_001274430.1 | NP_001261359.1 | 3 - 4 | 1506 | 95 | ||
| NM_001274431.1 | NP_001261360.1 | 3 - 4 | 1506 | 95 | ||
| NM_139523.3 | NP_647780.1 | 3 - 4 | 1506 | 95 | ||
| NM_206262.3 | NP_995984.1 | 3 - 4 | 1506 | 95 | ||
| GenBank mRNA | AY094827.1 | - | 3 - 4 | 1502 | 95 | |
More Information
Set Membership: |
|

Girdin
Genomic Map