Assay Details
| Gene Symbol: FASN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144306.2 | NP_001137778.2 | 4 - 5 | 7491 | 86 | |
| NM_134904.3 | NP_608748.1 | 4 - 5 | 7696 | 86 | ||
More Information
Set Membership: |
|

FASN1
Genomic Map