Assay Details
| Gene Symbol: Cpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273199.1 | NP_001260128.1 | 4 - 5 | 792 | 77 | |
| NM_001273200.1 | NP_001260129.1 | 4 - 5 | 890 | 77 | ||
| NM_057810.4 | NP_477158.1 | 4 - 5 | 950 | 77 | ||
| NM_164689.2 | NP_723173.1 | 2 - 3 | 368 | 77 | ||
| GenBank mRNA | AY052000.1 | - | 4 - 5 | 930 | 77 | |
More Information
Set Membership: |
|

Cpr
Genomic Map