Assay Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135703.4 | NP_609547.2 | 4 - 5 | 741 | 61 | |
| NM_164991.3 | NP_723722.2 | 5 - 6 | 1120 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pex19| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135703.4 | NP_609547.2 | 4 - 5 | 741 | 61 | |
| NM_164991.3 | NP_723722.2 | 5 - 6 | 1120 | 61 | ||
Set Membership: |
|