Assay Details
| Gene Symbol: hep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272571.2 | NP_001259500.1 | 4 - 5 | 1970 | 74 | |
| NM_001272572.2 | NP_001259501.1 | 4 - 5 | 1970 | 74 | ||
| NM_001298253.1 | NP_001285182.1 | 4 - 5 | 1970 | 74 | ||
| NM_001382110.1 | NP_001368981.1 | 4 - 5 | 1970 | 74 | ||
| NM_001382111.1 | NP_001368982.1 | 4 - 5 | 1970 | 74 | ||
| NM_078587.3 | NP_511142.2 | 4 - 5 | 1970 | 74 | ||
| NM_167346.2 | NP_727661.1 | 4 - 5 | 1970 | 74 | ||
| GenBank mRNA | AY069695.1 | - | 4 - 5 | 1905 | 74 | |
| U93032.1 | - | 4 - 5 | 1325 | 74 | ||
More Information
Set Membership: |
|

hep
Genomic Map