Assay Details
| Gene Symbol: CycD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078631.4 | NP_523355.2 | 5 - 6 | 1375 | 68 | |
| NM_167472.3 | NP_727912.2 | 5 - 6 | 1486 | 68 | ||
| NM_167473.2 | NP_727913.2 | 4 - 5 | 1683 | 68 | ||
More Information
Set Membership: |
|

CycD
Genomic Map