Assay Details
| Gene Symbol: Mpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078885.3 | NP_523609.3 | 2 | 1095 | 124 | |
| GenBank mRNA | AY029612.1 | - | 1 | 563 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpp6| Gene Symbol: Mpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078885.3 | NP_523609.3 | 2 | 1095 | 124 | |
| GenBank mRNA | AY029612.1 | - | 1 | 563 | 124 | |
Set Membership: |
|