Assay Details
| Gene Symbol: Pink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031878.2 | NP_001027049.1 | 4 - 5 | 1574 | 165 | |
| NM_001031879.3 | NP_001027050.1 | 3 - 4 | 1601 | 165 | ||
| NM_001031880.3 | NP_001027051.1 | 4 - 5 | 1550 | 165 | ||
| NM_001031881.3 | NP_001027052.1 | 4 - 5 | 1544 | 165 | ||
| NM_001031882.3 | NP_001027053.1 | 4 - 5 | 1453 | 165 | ||
| NM_001031883.3 | NP_001027054.1 | 4 - 5 | 1545 | 165 | ||
| NM_001298023.1 | NP_001284952.1 | 3 - 4 | 1087 | 165 | ||
| NM_132112.5 | NP_572340.2 | 4 - 5 | 1539 | 165 | ||
| NM_167083.4 | NP_727110.1 | 4 - 5 | 1459 | 165 | ||
| GenBank mRNA | BT133188.1 | - | 4 - 5 | 1537 | 165 | |
More Information
Set Membership: |
|

Pink1
Genomic Map