Assay Details
| Gene Symbol: ci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258472.2 | NP_001245401.1 | 5 - 6 | 2119 | 66 | |
| NM_001258473.1 | NP_001245402.1 | 5 - 6 | 2436 | 66 | ||
| NM_079878.5 | NP_524617.3 | 5 - 6 | 2186 | 66 | ||
| GenBank mRNA | BT133159.1 | - | 5 - 6 | 2115 | 66 | |
More Information
Set Membership: |
|

ci
Genomic Map