Assay Details
| Gene Symbol: sip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298707.1 | NP_001285636.1 | 1 - 2 | 191 | 58 | |
| NM_079986.4 | NP_524725.2 | 1 - 2 | 191 | 58 | ||
| GenBank mRNA | AF145670.1 | - | 1 - 2 | 168 | 58 | |
More Information
Set Membership: |
|

sip1
Genomic Map