Assay Details
| Gene Symbol: Drsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168019.4 | NP_728860.2 | 1 | 87 | 59 | |
| GenBank mRNA | AY338362.1 | - | 1 | 61 | 59 | |
| AY351397.1 | - | 1 | 61 | 59 | ||
| EU375839.1 | - | 1 | 61 | 59 | ||
More Information
Set Membership: |
|

Drsl2
Genomic Map