Assay Details
| Gene Symbol: sli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103863.3 | NP_001097333.1 | 19 - 20 | 5517 | 65 | |
| NM_001103864.3 | NP_001097334.1 | 19 - 20 | 4746 | 65 | ||
| NM_001274088.1 | NP_001261017.1 | 20 - 21 | 5589 | 65 | ||
| NM_057379.4 | NP_476727.1 | 20 - 21 | 5589 | 65 | ||
| NM_057380.4 | NP_476728.1 | 19 - 20 | 5517 | 65 | ||
| NM_057381.4 | NP_476729.1 | 20 - 21 | 5546 | 65 | ||
More Information
Set Membership: |
|

sli
Genomic Map