Assay Details
| Gene Symbol: sli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103863.3 | NP_001097333.1 | 3 - 4 | 1539 | 71 | |
| NM_001103864.3 | NP_001097334.1 | 2 - 3 | 696 | 71 | ||
| NM_001274088.1 | NP_001261017.1 | 3 - 4 | 1539 | 71 | ||
| NM_057379.4 | NP_476727.1 | 3 - 4 | 1539 | 71 | ||
| NM_057380.4 | NP_476728.1 | 3 - 4 | 1539 | 71 | ||
| NM_057381.4 | NP_476729.1 | 4 - 5 | 1601 | 71 | ||
More Information
Set Membership: |
|

sli
Genomic Map