Assay Details
| Gene Symbol: Cbp53E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299582.1 | NP_001286511.1 | 10 - 11 | 1034 | 117 | |
| NM_057490.4 | NP_476838.1 | 9 - 10 | 985 | 117 | ||
| GenBank mRNA | AY058340.1 | - | 9 - 10 | 952 | 117 | |
| X68566.1 | - | 934 | 117 | |||
More Information
Set Membership: |
|

Cbp53E
Genomic Map