Assay Details
| Gene Symbol: CG14220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258830.2 | NP_001245759.1 | 1 - 2 | 245 | 130 | |
| NM_134481.2 | NP_608325.1 | 1 - 2 | 245 | 130 | ||
| GenBank mRNA | BT044333.1 | - | 1 - 2 | 244 | 130 | |
More Information
Set Membership: |
|

CG14220
Genomic Map