Assay Details
| Gene Symbol: TAF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275521.1 | NP_001262450.1 | 2 - 3 | 768 | 94 | |
| NM_001275522.1 | NP_001262451.1 | 2 - 3 | 663 | 94 | ||
| NM_141725.3 | NP_649982.1 | 2 - 3 | 663 | 94 | ||
| GenBank mRNA | BT057993.1 | - | 2 - 3 | 653 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001275521.1 transcripts.
Set Membership: |
|

TAF1B
Genomic Map