Assay Details
| Gene Symbol: HisCl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170120.2 | NP_001163591.1 | 5 - 6 | 1037 | 97 | |
| NM_141859.4 | NP_650116.2 | 5 - 6 | 1055 | 97 | ||
| NM_169429.4 | NP_731632.1 | 5 - 6 | 1055 | 97 | ||
| NM_206474.3 | NP_996196.1 | 3 - 4 | 1051 | 97 | ||
| GenBank mRNA | AF382401.1 | - | 4 - 5 | 937 | 97 | |
| AF382402.1 | - | 4 - 5 | 937 | 97 | ||
| AF435470.1 | - | 4 - 5 | 739 | 97 | ||
| AF435471.1 | - | 3 - 4 | 853 | 97 | ||
| AY049775.1 | - | 4 - 5 | 891 | 97 | ||
| AY060303.1 | - | 3 - 4 | 1005 | 97 | ||
More Information
Set Membership: |
|

HisCl1
Genomic Map