Assay Details
| Gene Symbol: VhaPPA1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142149.3 | NP_650406.2 | 1 | 372 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VhaPPA1-2| Gene Symbol: VhaPPA1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142149.3 | NP_650406.2 | 1 | 372 | 74 | |
Set Membership: |
|