Assay Details
| Gene Symbol: Lrrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104377.2 | NP_001097847.1 | 3 - 4 | 1590 | 69 | |
| NM_001275842.1 | NP_001262771.1 | 3 - 4 | 1590 | 69 | ||
| NM_001275843.1 | NP_001262772.1 | 4 - 5 | 1863 | 69 | ||
| GenBank mRNA | DQ494533.1 | - | 3 - 4 | 1501 | 69 | |
More Information
Set Membership: |
|

Lrrk
Genomic Map