Assay Details
| Gene Symbol: Nmnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276013.2 | NP_001262942.1 | 4 - 5 | 846 | 80 | |
| NM_143058.1 | NP_651315.2 | 4 - 5 | 882 | 80 | ||
| GenBank mRNA | BT030750.1 | - | 4 - 5 | 881 | 80 | |
More Information
Set Membership: |
|

Nmnat
Genomic Map