Assay Details
| Gene Symbol: Nmnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001369970.1 | NP_001356924.1 | 4 - 5 | 844 | 70 | |
| NM_170185.1 | NP_733064.2 | 4 - 5 | 880 | 70 | ||
| GenBank mRNA | AY089411.1 | - | 4 - 5 | 807 | 70 | |
More Information
Set Membership: |
|

Nmnat
Genomic Map