Assay Details
| Gene Symbol: Mgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014684.2 | NP_001014684.1 | 8 - 9 | 1591 | 58 | |
| NM_001276167.1 | NP_001263096.1 | 9 - 10 | 1953 | 58 | ||
| NM_143506.4 | NP_651763.4 | 9 - 10 | 2250 | 58 | ||
| GenBank mRNA | AB176553.1 | - | 8 - 9 | 1605 | 58 | |
| AY047566.1 | - | 9 - 10 | 2216 | 58 | ||
More Information
Set Membership: |
|

Mgat2
Genomic Map