Assay Details
| Gene Symbol: Mgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014684.2 | NP_001014684.1 | 5 - 6 | 1064 | 58 | |
| NM_001276167.1 | NP_001263096.1 | 5 - 6 | 1363 | 58 | ||
| NM_143506.4 | NP_651763.4 | 5 - 6 | 1363 | 58 | ||
| GenBank mRNA | AB176553.1 | - | 4 - 5 | 718 | 58 | |
| AY047566.1 | - | 5 - 6 | 1329 | 58 | ||
More Information
Set Membership: |
|

Mgat2
Genomic Map