Assay Details
| Gene Symbol: Cpr73D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104157.3 | NP_001097627.1 | 4 | 2104 | 96 | |
| NM_001316452.1 | NP_001303381.1 | 5 | 2622 | 96 | ||
| GenBank mRNA | AY051482.1 | - | 3 | 1651 | 96 | |
More Information
Set Membership: |
|

Cpr73D
Genomic Map