Assay Details
| Gene Symbol: Dyrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038721.2 | NP_001033810.1 | 6 - 7 | 2669 | 105 | |
| NM_001038723.2 | NP_001033812.1 | 5 - 6 | 2605 | 105 | ||
| NM_001038725.2 | NP_001033814.1 | 6 - 7 | 2669 | 105 | ||
| GenBank mRNA | AY118401.1 | - | 6 - 7 | 2663 | 105 | |
More Information
Set Membership: |
|

Dyrk3
Genomic Map