Assay Details
| Gene Symbol: mzt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316934.1 | NP_001303863.1 | 3 - 4 | 82 | ||
| GenBank mRNA | BC078275.1 | - | 3 - 4 | 405 | 82 | |
| CK396487.1 | - | 3 - 4 | 82 | |||
More Information
Set Membership: |
|

mzt2b
Genomic Map