Assay Details
| Gene Symbol: ndufa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013456.1 | NP_001013474.1 | 2 - 3 | 73 | 69 | |
| GenBank mRNA | BC091561.1 | - | 2 - 3 | 73 | 69 | |
More Information
Set Membership: |
|

ndufa5
Genomic Map