Assay Details
| Gene Symbol: phf23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013499.1 | NP_001013517.1 | 3 - 4 | 665 | 108 | |
| XM_021476141.1 | XP_021331816.1 | 3 - 4 | 311 | 108 | ||
| GenBank mRNA | BC091667.1 | - | 3 - 4 | 665 | 108 | |
| BC153421.1 | - | 4 - 5 | 835 | 108 | ||
| BC163957.1 | - | 3 - 4 | 188 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021476141.1 transcripts.
Set Membership: |
|

phf23a
Genomic Map