Assay Details
| Gene Symbol: ubr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212629.1 | NP_997794.1 | 8 - 9 | 1096 | 64 | |
| XM_021468365.1 | XP_021324040.1 | 8 - 9 | 1502 | 64 | ||
| GenBank mRNA | BC044390.1 | - | 8 - 9 | 1096 | 64 | |
More Information
Set Membership: |
|

ubr7
Genomic Map