Assay Details
| Gene Symbol: mpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351837.1 | NP_001338766.1 | 6 - 7 | 659 | 72 | |
| NM_212779.2 | NP_997944.2 | 6 - 7 | 659 | 72 | ||
| XM_021479113.1 | XP_021334788.1 | 6 - 7 | 722 | 72 | ||
| GenBank mRNA | AF349034.1 | - | 6 - 7 | 660 | 72 | |
| AF378824.1 | - | 3 - 4 | 336 | 72 | ||
| BC068379.1 | - | 6 - 7 | 634 | 72 | ||
More Information
Set Membership: |
|

mpx
Genomic Map