Assay Details
| Gene Symbol: dnajb6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002353.1 | NP_001002353.1 | 2 - 3 | 137 | 72 | |
| XM_005163424.4 | XP_005163481.1 | 2 - 3 | 190 | 72 | ||
| XM_005163425.4 | XP_005163482.1 | 2 - 3 | 228 | 72 | ||
| GenBank mRNA | BC075905.1 | - | 2 - 3 | 137 | 72 | |
More Information
Set Membership: |
|

dnajb6a
Genomic Map