Assay Details
| Gene Symbol: ccdc106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012490.2 | NP_001012508.1 | 5 - 6 | 563 | 94 | |
| GenBank mRNA | BC090178.1 | - | 5 - 6 | 568 | 94 | |
More Information
Set Membership: |
|

ccdc106a
Genomic Map