Assay Details
| Gene Symbol: usf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213194.1 | NP_998359.1 | 5 - 6 | 402 | 72 | |
| XM_005171828.4 | XP_005171885.1 | 5 - 6 | 445 | 72 | ||
| GenBank mRNA | BC065876.1 | - | 5 - 6 | 402 | 72 | |
More Information
Set Membership: |
|

usf1l
Genomic Map