Assay Details
| Gene Symbol: pacsin1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328357.1 | NP_001315286.1 | 3 - 4 | 440 | 56 | |
| XM_005161905.4 | XP_005161962.1 | 3 - 4 | 420 | 56 | ||
| XM_021469807.1 | XP_021325482.1 | 3 - 4 | 421 | 56 | ||
| GenBank mRNA | BC095091.1 | - | 3 - 4 | 408 | 56 | |
| BC164592.1 | - | 2 - 3 | 255 | 56 | ||
More Information
Set Membership: |
|

pacsin1a
Genomic Map