Assay Details
| Gene Symbol: dcaf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077552.1 | NP_001071020.1 | 2 - 3 | 524 | 73 | |
| GenBank mRNA | BC124494.1 | - | 2 - 3 | 524 | 73 | |
More Information
Set Membership: |
|

dcaf5
Genomic Map