Assay Details
| Gene Symbol: slc4a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198338.1 | NP_938152.1 | 13 - 14 | 1495 | 108 | |
| GenBank mRNA | AF350072.1 | - | 13 - 14 | 1495 | 108 | |
| AY098936.1 | - | 13 - 14 | 1528 | 108 | ||
| BC093126.1 | - | 6 - 7 | 796 | 108 | ||
| BC163932.1 | - | 13 - 14 | 1481 | 108 | ||
More Information
Set Membership: |
|

slc4a1a
Genomic Map