Assay Details
| Gene Symbol: naa15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200646.2 | NP_956940.2 | 2 - 3 | 341 | 119 | |
| GenBank mRNA | BC045491.1 | - | 2 - 3 | 310 | 119 | |
| BC057466.1 | - | 2 - 3 | 119 | |||
| BC165243.1 | - | 2 - 3 | 174 | 119 | ||
More Information
Set Membership: |
|

naa15a
Genomic Map