Assay Details
| Gene Symbol: acat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131370.2 | NP_571445.2 | 7 - 8 | 984 | 110 | |
| XM_021468355.1 | XP_021324030.1 | 6 - 7 | 110 | |||
| GenBank mRNA | AF143488.1 | - | 7 - 8 | 110 | ||
| BC045949.1 | - | 7 - 8 | 984 | 110 | ||
| BC165310.1 | - | 7 - 8 | 941 | 110 | ||
More Information
Set Membership: |
|

acat2
Genomic Map