Assay Details
| Gene Symbol: amy2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213011.2 | NP_998176.2 | 7 - 8 | 1126 | 57 | |
| GenBank mRNA | BC054669.1 | - | 7 - 8 | 1101 | 57 | |
| BC062867.1 | - | 7 - 8 | 1103 | 57 | ||
More Information
Set Membership: |
|

amy2a
Genomic Map