Assay Details
| Gene Symbol: znf143a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007441.1 | NP_001007442.1 | 12 - 13 | 1516 | 92 | |
| GenBank mRNA | BC085401.1 | - | 12 - 13 | 1516 | 92 | |
| BC165773.1 | - | 11 - 12 | 1422 | 92 | ||
More Information
Set Membership: |
|

znf143a
Genomic Map