Assay Details
| Gene Symbol: mocos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014366.2 | NP_001014388.2 | 1 - 2 | 145 | 85 | |
| XM_009307222.3 | XP_009305497.1 | 1 - 2 | 249 | 85 | ||
| GenBank mRNA | BC091876.1 | - | 1 - 2 | 147 | 85 | |
| BC164847.1 | - | 1 - 2 | 158 | 85 | ||
| CK681135.1 | - | 1 - 2 | 147 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009307222.3 transcripts.
Set Membership: |
|

mocos
Genomic Map