Assay Details
| Gene Symbol: ccdc125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098182.1 | NP_001091652.1 | 1 - 2 | 237 | 62 | |
| XM_017356141.2 | XP_017211630.1 | 3 - 4 | 412 | 62 | ||
| GenBank mRNA | BC163381.1 | - | 1 - 2 | 237 | 62 | |
| BC163384.1 | - | 1 - 2 | 237 | 62 | ||
More Information
Set Membership: |
|

ccdc125
Genomic Map