Assay Details
| Gene Symbol: klhl41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198979.1 | NP_945330.1 | 2 - 3 | 1367 | 72 | |
| GenBank mRNA | BC054912.1 | - | 2 - 3 | 1367 | 72 | |
More Information
Set Membership: |
|

klhl41b
Genomic Map