Assay Details
| Gene Symbol: gfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077746.2 | NP_001071214.2 | 3 - 4 | 831 | 60 | |
| GenBank mRNA | BC125879.1 | - | 3 - 4 | 768 | 60 | |
More Information
Set Membership: |
|

gfod2
Genomic Map