Assay Details
| Gene Symbol: smarca4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181603.1 | NP_853634.1 | 19 - 20 | 3010 | 84 | |
| XM_021469728.1 | XP_021325403.1 | 20 - 21 | 3197 | 84 | ||
| GenBank mRNA | AY205256.1 | - | 19 - 20 | 3010 | 84 | |
| BC163918.1 | - | 19 - 20 | 3072 | 84 | ||
| BC163920.1 | - | 19 - 20 | 3072 | 84 | ||
More Information
Set Membership: |
|

smarca4a
Genomic Map