Assay Details
| Gene Symbol: lipca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201022.1 | NP_957316.1 | 6 - 7 | 1108 | 64 | |
| GenBank mRNA | BC053243.1 | - | 6 - 7 | 1108 | 64 | |
| BC163963.1 | - | 7 - 8 | 1083 | 64 | ||
More Information
Set Membership: |
|

lipca
Genomic Map