Assay Details
| Gene Symbol: rnf175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328343.1 | NP_001315272.1 | 7 - 8 | 802 | 76 | |
| NM_200308.3 | NP_956602.2 | 8 - 9 | 839 | 76 | ||
| GenBank mRNA | BC050174.1 | - | 8 - 9 | 840 | 76 | |
| BC071470.1 | - | 7 - 8 | 793 | 76 | ||
More Information
Set Membership: |
|

rnf175
Genomic Map