Assay Details
| Gene Symbol: tfap2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025546.1 | NP_001020717.1 | 7 - 8 | 1362 | 67 | |
| XM_005160887.4 | XP_005160944.1 | 7 - 8 | 1640 | 67 | ||
| GenBank mRNA | BC097244.1 | - | 7 - 8 | 1362 | 67 | |
More Information
Set Membership: |
|

tfap2d
Genomic Map